Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008779.hg.1 |
TC0X000666.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008779.hg.1 |
ENST00000370629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008779.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158144.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158144.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158144.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158144.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158144.hg.1 |
TC0X00008540.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158143.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158143.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158143.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158143.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158143.hg.1 |
TC0X00008540.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
ENST00000370629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008540.hg.1 |
NM_000074.2 |
Refseq |