Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006297.2 | NP_006288.2 | 4 | 524 | 73 | |
GenBank mRNA | AB208781.1 | - | 4 | 447 | 73 | |
AK293542.1 | - | 4 | 502 | 73 | ||
AK300163.1 | - | 3 | 416 | 73 | ||
AK315332.1 | - | 4 | 513 | 73 | ||
BC023593.2 | - | 4 | 411 | 4646806 | 73 | |
BP349667.1 | - | 4 | 524 | 73 | ||
CR456728.1 | - | 4 | 404 | 73 | ||
M36089.1 | - | 4 | 952 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons |