Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545442.2 | XP_005545499.1 | 1 - 2 | 267 | 86 | |
GenBank mRNA | M19318.1 | - | 1 - 2 | 249 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |