Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565984.2 | XP_005566041.1 | 1 - 2 | 253 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |