Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594158.2 | XP_005594215.1 | 1 - 2 | 305 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |