Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284730.1 | NP_001271659.1 | 3 - 4 | 576 | 109 | |
XM_005572570.1 | XP_005572627.1 | 4 - 5 | 630 | 109 | ||
GenBank mRNA | AB169613.1 | - | 3 - 4 | 576 | 109 | |
AB171798.1 | - | 3 - 4 | 576 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |