Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102115404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579334.1 | XP_005579391.1 | 3 - 4 | 834 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |