Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593944.2 | XP_005594001.1 | 2 - 3 | 558 | 75 | |
XM_005593945.2 | XP_005594002.1 | 2 - 3 | 261 | 75 | ||
XM_005593946.2 | XP_005594003.1 | 2 - 3 | 1268 | 75 | ||
XM_005593947.2 | XP_005594004.1 | 2 - 3 | 283 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |