Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567205.2 | XP_005567262.1 | 1 - 2 | 570 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |