Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557229.2 | XP_005557286.1 | 2 - 3 | 259 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |