Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563345.2 | XP_005563402.1 | 2 - 3 | 294 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |