Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591275.2 | XP_005591332.1 | 3 | 293 | 112 | |
XM_005591276.2 | XP_005591333.2 | 2 | 4320 | 112 | ||
XM_015442403.1 | XP_015297889.1 | 3 | 4353 | 112 | ||
XM_015442404.1 | XP_015297890.1 | 4 | 1585 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |