Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578156.2 | XP_005578213.1 | 4 | 2710 | 113 | |
XM_005578158.2 | XP_005578215.1 | 2 | 2588 | 113 | ||
XM_015434892.1 | XP_015290378.1 | 12 | 3029 | 113 | ||
XM_015434893.1 | XP_015290379.1 | 12 | 3841 | 113 | ||
XM_015434894.1 | XP_015290380.1 | 11 | 3688 | 113 | ||
XM_015434895.1 | XP_015290381.1 | 10 | 3634 | 113 | ||
XM_015434896.1 | XP_015290382.1 | 11 | 2977 | 113 | ||
XM_015434897.1 | XP_015290383.1 | 11 | 2887 | 113 | ||
XM_015434898.1 | XP_015290384.1 | 11 | 2968 | 113 | ||
XM_015434899.1 | XP_015290385.1 | 10 | 3873 | 113 | ||
XM_015434900.1 | XP_015290386.1 | 9 | 3429 | 113 | ||
XM_015434901.1 | XP_015290387.1 | 6 | 2842 | 113 | ||
XM_015434902.1 | XP_015290388.1 | 8 | 4828 | 113 | ||
XM_015434903.1 | XP_015290389.1 | 5 | 4194 | 113 | ||
XM_015434904.1 | XP_015290390.1 | 5 | 2762 | 113 | ||
XM_015434905.1 | XP_015290391.1 | 5 | 7845 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |