Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578093.2 | XP_005578150.1 | 1 - 2 | 1929 | 98 | |
XM_005578096.2 | XP_005578153.1 | 2 - 3 | 1046 | 98 | ||
XM_005578097.2 | XP_005578154.1 | 2 - 3 | 949 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |