Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015450814.1 | XP_015306300.1 | 9 - 10 | 1055 | 67 | |
XM_015450815.1 | XP_015306301.1 | 8 - 9 | 943 | 67 | ||
XM_015450816.1 | XP_015306302.1 | 8 - 9 | 1449 | 67 | ||
XM_015450817.1 | XP_015306303.1 | 9 - 10 | 1055 | 67 | ||
XM_015450818.1 | XP_015306304.1 | 7 - 8 | 881 | 67 | ||
XM_015450819.1 | XP_015306305.1 | 6 - 7 | 863 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |