Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546393.2 | XP_005546450.1 | 1 - 2 | 2874 | 63 | |
GenBank mRNA | AY675630.1 | - | 1 - 2 | 774 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Probe spans exons |