Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593002.1 | XP_005593059.1 | 1 - 2 | 138 | 93 | |
XM_005593003.2 | XP_005593060.1 | 1 - 2 | 140 | 93 | ||
XM_015443621.1 | XP_015299107.1 | 1 - 2 | 131 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |