Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579132.2 | XP_005579189.1 | 6 - 7 | 705 | 69 | |
XM_005579134.2 | XP_005579191.1 | 5 - 6 | 586 | 69 | ||
XM_005579135.2 | XP_005579192.1 | 6 - 7 | 705 | 69 | ||
XM_005579138.2 | XP_005579195.1 | 6 - 7 | 705 | 69 | ||
XM_015435511.1 | XP_015290997.1 | 6 - 7 | 705 | 69 | ||
XM_015435512.1 | XP_015290998.1 | 5 - 6 | 586 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |