Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555051.2 | XP_005555108.1 | 2 - 3 | 196 | 112 | |
XM_005555052.2 | XP_005555109.1 | 2 - 3 | 623 | 112 | ||
XM_005555053.2 | XP_005555110.1 | 2 - 3 | 281 | 112 | ||
XM_005555054.2 | XP_005555111.1 | 2 - 3 | 189 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |