Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575302.2 | XP_005575359.1 | 1 - 2 | 82 | 109 | |
XM_015433400.1 | XP_015288886.1 | 1 - 2 | 82 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |