Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582566.2 | XP_005582623.1 | 2 - 3 | 1012 | 90 | |
XM_015437475.1 | XP_015292961.1 | 2 - 3 | 1015 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |