Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583561.2 | XP_005583618.1 | 1 - 2 | 661 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |