Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588956.2 | XP_005589013.1 | 2 - 3 | 158 | 108 | |
XM_005588957.2 | XP_005589014.1 | 2 - 3 | 256 | 108 | ||
XM_005588958.2 | XP_005589015.1 | 2 - 3 | 158 | 108 | ||
XM_015441758.1 | XP_015297244.1 | 1 - 2 | 110 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |