Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015444869.1 | XP_015300355.1 | 3 - 4 | 853 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |