Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102138242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442800.1 | XP_015298286.1 | 2 - 3 | 325 | 51 | |
XM_015442801.1 | XP_015298287.1 | 2 - 3 | 325 | 51 | ||
GenBank mRNA | AB060195.1 | - | 2 - 3 | 282 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |