Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547493.2 | XP_005547550.1 | 3 | 2974 | 140 | |
XM_005547494.2 | XP_005547551.1 | 3 | 1042 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |