Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545139.2 | XP_005545196.1 | 2 | 771 | 140 | |
XM_005545140.2 | XP_005545197.1 | 2 | 466 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |