Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283438.1 | NP_001270367.1 | 3 - 4 | 1123 | 85 | |
GenBank mRNA | AF104339.1 | - | 3 - 4 | 1123 | 85 |
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350029.1 | NP_001336958.1 | 3 - 4 | 1089 | 85 |
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350028.1 | NP_001336957.1 | 3 - 4 | 1179 | 85 | |
GenBank mRNA | AF104337.1 | - | 3 - 4 | 1180 | 85 |
Gene Symbol: UGT1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350023.1 | NP_001336952.1 | 3 - 4 | 1102 | 85 | |
GenBank mRNA | AF104338.1 | - | 3 - 4 | 1103 | 85 |
Gene Symbol: UGT1A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350026.1 | NP_001336955.1 | 3 - 4 | 1136 | 85 | |
GenBank mRNA | AF104336.1 | - | 3 - 4 | 1136 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |