Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285277.1 | NP_001272206.1 | 2 - 3 | 258 | 82 | |
XM_005553562.2 | XP_005553619.1 | 1 - 2 | 1014 | 82 | ||
GenBank mRNA | AB000513.1 | - | 2 - 3 | 258 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |