Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567263.2 | XP_005567320.1 | 3 - 4 | 423 | 90 | |
XM_005567264.2 | XP_005567321.1 | 3 - 4 | 390 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |