Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284145.1 | NP_001271074.1 | 3 - 4 | 359 | 76 | |
GenBank mRNA | DQ078723.1 | - | 3 - 4 | 359 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |