Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005562014.2 | XP_005562071.1 | 1 - 2 | 275 | 80 | |
GenBank mRNA | AB220420.1 | - | 1 - 2 | 205 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |