Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564053.2 | XP_005564110.1 | 2 - 3 | 286 | 80 | |
XM_005564054.2 | XP_005564111.1 | 3 - 4 | 392 | 80 | ||
XM_005564055.2 | XP_005564112.1 | 2 - 3 | 247 | 80 | ||
XM_015455167.1 | XP_015310653.1 | 3 - 4 | 311 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |