Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593568.2 | XP_005593625.1 | 4 - 5 | 552 | 116 | |
XM_005593569.2 | XP_005593626.1 | 4 - 5 | 361 | 116 | ||
XM_005593570.2 | XP_005593627.1 | 4 - 5 | 381 | 116 | ||
XM_005593571.2 | XP_005593628.1 | 4 - 5 | 431 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |