Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587577.2 | XP_005587634.2 | 1 - 2 | 109 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |