Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555218.2 | XP_005555275.1 | 1 - 2 | 145 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |