Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015456458.1 | XP_015311944.1 | 11 - 12 | 1751 | 139 | |
XM_015456459.1 | XP_015311945.1 | 8 - 9 | 3091 | 139 | ||
XM_015456460.1 | XP_015311946.1 | 11 - 12 | 1751 | 139 | ||
XM_015456461.1 | XP_015311947.1 | 11 - 12 | 1751 | 139 | ||
XM_015456462.1 | XP_015311948.1 | 11 - 12 | 1751 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |