Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554497.2 | XP_005554554.1 | 1 - 2 | 316 | 74 | |
XM_015450180.1 | XP_015305666.1 | 2 - 3 | 304 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |