Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574854.2 | XP_005574911.1 | 1 - 2 | 493 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |