Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287668.1 | NP_001274597.1 | 6 - 7 | 529 | 84 | |
XM_005592529.2 | XP_005592586.1 | 6 - 7 | 1112 | 84 | ||
XM_005592532.2 | XP_005592589.1 | 4 - 5 | 533 | 84 | ||
XM_015442716.1 | XP_015298202.1 | 5 - 6 | 577 | 84 | ||
XM_015442717.1 | XP_015298203.1 | 6 - 7 | 685 | 84 | ||
XM_015442718.1 | XP_015298204.1 | 5 - 6 | 631 | 84 | ||
GenBank mRNA | AB179265.1 | - | 6 - 7 | 529 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |