Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577517.2 | XP_005577574.1 | 3 - 4 | 246 | 83 | |
XM_015434582.1 | XP_015290068.1 | 2 - 3 | 218 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Inventoried Probe spans exons |