Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541317.2 | XP_005541374.1 | 4 - 5 | 1101 | 93 | |
XM_005541318.2 | XP_005541375.1 | 4 - 5 | 1101 | 93 | ||
XM_015455099.1 | XP_015310585.1 | 6 - 7 | 1602 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Inventoried Probe spans exons |