Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541239.2 | XP_005541296.1 | 3 - 4 | 730 | 64 | |
XM_005541240.2 | XP_005541297.1 | 3 - 4 | 766 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Inventoried Probe spans exons |