Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589957.2 | XP_005590014.1 | 3 - 4 | 1162 | 132 | |
XM_005589958.2 | XP_005590015.1 | 3 - 4 | 1181 | 132 | ||
XM_015441178.1 | XP_015296664.1 | 3 - 4 | 3553 | 132 | ||
XM_015441180.1 | XP_015296666.1 | 3 - 4 | 613 | 132 | ||
XM_015441181.1 | XP_015296667.1 | 3 - 4 | 566 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |