Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284538.1 | NP_001271467.1 | 2 - 3 | 115 | 61 | |
GenBank mRNA | AB172479.1 | - | 2 - 3 | 115 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |