Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591761.2 | XP_005591818.1 | 1 - 2 | 322 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |