Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555877.2 | XP_005555934.1 | 3 - 4 | 1786 | 115 | |
XM_005555878.2 | XP_005555935.1 | 2 - 3 | 6515 | 115 | ||
XM_015450807.1 | XP_015306293.1 | 2 - 3 | 2486 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |