Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547770.2 | XP_005547827.1 | 1 - 2 | 256 | 82 | |
GenBank mRNA | AB168189.1 | - | 169 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |