Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574597.2 | XP_005574654.1 | 2 | 1017 | 129 | |
XM_015432951.1 | XP_015288437.1 | 2 | 2011 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |