Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARBI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578036.2 | XP_005578093.1 | 2 - 3 | 953 | 118 | |
XM_005578037.2 | XP_005578094.1 | 2 - 3 | 1190 | 118 | ||
XM_005578038.2 | XP_005578095.1 | 2 - 3 | 883 | 118 | ||
XM_005578039.2 | XP_005578096.1 | 2 - 3 | 1210 | 118 | ||
XM_005578040.2 | XP_005578097.1 | 2 - 3 | 968 | 118 | ||
XM_005578041.2 | XP_005578098.1 | 2 - 3 | 937 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |